(5-Chloro-4-methylthiophen-2-yl)triethylsilane ≥97%

Supplier: AOBChem USA

Warning

415031-5G 415031-1G 415031-25G 415031-500MG
77629-122EA 1484.23 USD
77629-122 77638-608 77659-196 77650-610
(5-Chloro-4-methylthiophen-2-yl)triethylsilane ≥97%
(5-Chloro-4-methylthiophen-2-yl)triethylsilane

Precaución: For Research Use Only

Formula: 246.872 g/mol
MW: C₁₁H₁₉ClSSi
Storage Temperature: Ambient
CAS Number: 3031808-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR