1-(5-Chloro-4-methylthiophen-2-yl)-2-ethylbutan-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
415029-5G
415029-25G
415029-1G
415029-500MG
77629-120EA
1484.23
USD
77629-120
77635-780
77642-642
77646-676
1-(5-Chloro-4-methylthiophen-2-yl)-2-ethylbutan-1-ol ≥95%
1-(5-Chloro-4-methylthiophen-2-yl)-2-ethylbutan-1-ol
Precaución: For Research Use Only
Formula:
232.77 g/mol MW: C₁₁H₁₇ClOS Storage Temperature: Ambient |
CAS Number:
1598484-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...