1-(5-Chloro-4-methylthiophen-2-yl)-2,2-dimethylpropan-1-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
415028-5G
415028-1G
415028-500MG
77629-118EA
1484.23
USD
77629-118
77642-640
77646-674
1-(5-Chloro-4-methylthiophen-2-yl)-2,2-dimethylpropan-1-ol ≥97%
1-(5-Chloro-4-methylthiophen-2-yl)-2,2-dimethylpropan-1-ol
Precaución: For Research Use Only
Formula:
218.744 g/mol MW: C₁₀H₁₅ClOS Storage Temperature: Ambient |
CAS Number:
1599325-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...