2-Chloro-1-(5-chloro-4-methylthiophen-2-yl)-2,2-difluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
415016-5G
415016-1G
415016-500MG
77629-114EA
1484.23
USD
77629-114
77638-606
77646-670
2-Chloro-1-(5-chloro-4-methylthiophen-2-yl)-2,2-difluoroethanone ≥97%
2-Chloro-1-(5-chloro-4-methylthiophen-2-yl)-2,2-difluoroethanone
Precaución: For Research Use Only
Formula:
245.074 g/mol MW: C₇H₄Cl₂F₂OS Storage Temperature: Ambient |
CAS Number:
3031813-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...