(5-Chloro-4-methylthiophen-2-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

415012-5G 415012-1G 415012-500MG
77629-112EA 1484.23 USD
77629-112 77638-604 77646-666
(5-Chloro-4-methylthiophen-2-yl)(phenyl)methanol ≥95%
(5-Chloro-4-methylthiophen-2-yl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 238.733 g/mol
MW: C₁₂H₁₁ClOS
Storage Temperature: Ambient
CAS Number: 1594454-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR