(5-Chloro-4-methylthiophen-2-yl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

415011-5G 415011-1G 415011-500MG
77629-110EA 1484.23 USD
77629-110 77642-634 77646-664
(5-Chloro-4-methylthiophen-2-yl)(cyclohexyl)methanol ≥95%
(5-Chloro-4-methylthiophen-2-yl)(cyclohexyl)methanol

Precaución: For Research Use Only

Formula: C₁₂H₁₇ClOS
Storage Temperature: Ambient
CAS Number: 1592343-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR