(5-Chloro-4-methylthiophen-2-yl)(cyclopropyl)methanol ≥97%

Supplier: AOBChem USA

Warning

415010-5G 415010-1G 415010-500MG
77655-612EA 2073 USD
77655-612 77642-632 77650-608
(5-Chloro-4-methylthiophen-2-yl)(cyclopropyl)methanol ≥97%
(5-Chloro-4-methylthiophen-2-yl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: 202.701 g/mol
MW: C₉H₁₁ClOS
Storage Temperature: Ambient
CAS Number: 1593029-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR