(5-Chloro-4-methylthiophen-2-yl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

415009-5G 415009-25G 415009-1G 415009-500MG
77633-074EA 1328 USD
77633-074 77655-610 77642-630 77646-662
(5-Chloro-4-methylthiophen-2-yl)(cyclopropyl)methanone ≥95%
(5-Chloro-4-methylthiophen-2-yl)(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: 200.685 g/mol
MW: C₉H₉ClOS
Storage Temperature: Ambient
CAS Number: 1592553-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR