1-(5-Chloro-4-methylthiophen-2-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

415007-5G 415007-1G 415007-500MG
77629-108EA 1484.23 USD
77629-108 77638-602 77646-660
1-(5-Chloro-4-methylthiophen-2-yl)-2,2-difluoroethanone ≥95%
1-(5-Chloro-4-methylthiophen-2-yl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: 210.629 g/mol
MW: C₇H₅ClF₂OS
Storage Temperature: Ambient
CAS Number: 1702137-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR