1-(5-Chloro-4-methylthiophen-2-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

415006-5G 415006-25G 415006-500MG 415006-1G
77629-106EA 1484.23 USD
77629-106 77635-778 77652-774 77642-628
1-(5-Chloro-4-methylthiophen-2-yl)-2,2,2-trifluoroethanone ≥95%
1-(5-Chloro-4-methylthiophen-2-yl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 228.619 g/mol
MW: C₇H₄ClF₃OS
Storage Temperature: Ambient
CAS Number: 1596974-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR