(5-Chloro-4-methylthiophen-2-yl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

415005-5G 415005-25G 415005-1G 415005-500MG
77633-072EA 1226 USD
77633-072 77655-608 77646-658 77650-606
(5-Chloro-4-methylthiophen-2-yl)trimethylsilane ≥95%
(5-Chloro-4-methylthiophen-2-yl)trimethylsilane

Precaución: For Research Use Only

Formula: C₈H₁₃ClSSi
Storage Temperature: Ambient
CAS Number: 3031812-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR