2-Fluoro-3-methyl-5-nitrobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

413984-5G 413984-1G 413984-500MG 413984-25G
77633-070EA 1296 USD
77633-070 77638-600 77652-772 77659-190
2-Fluoro-3-methyl-5-nitrobenzoic acid ≥97%
2-Fluoro-3-methyl-5-nitrobenzoic acid

Precaución: For Research Use Only

Formula: C₈H₆FNO₄
Storage Temperature: Ambient
CAS Number: 1519287-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR