Ethyl 3-fluoro-2-methoxy-α-oxo-4-pyridineacetate ≥97%

Supplier: AOBChem USA

Warning

412025-5G 412025-25G 412025-1G 412025-500MG
77629-102EA 1484.23 USD
77629-102 77635-774 77638-598 77650-602
Ethyl 3-fluoro-2-methoxy-α-oxo-4-pyridineacetate ≥97%
Ethyl 3-fluoro-2-methoxy-α-oxo-4-pyridineacetate

Precaución: For Research Use Only

Formula: 227.19 g/mol
MW: C₁₀H₁₀FNO₄
Storage Temperature: Ambient
CAS Number: 2000146-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR