Methyl 3-chloro-2-methoxy-α-oxo-4-pyridineacetate ≥95%

Supplier: AOBChem USA

Warning

412018-1G 412018-5G 412018-500MG
77638-596EA 676 USD
77638-596 77659-188 77650-600
Methyl 3-chloro-2-methoxy-α-oxo-4-pyridineacetate ≥95%
Methyl 3-chloro-2-methoxy-α-oxo-4-pyridineacetate

Precaución: For Research Use Only

Formula: 229.62 g/mol
MW: C₉H₈ClNO₄
Storage Temperature: Ambient
CAS Number: 2164169-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR