Methyl α,3-dihydroxybenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

410243-5G 410243-25G 410243-1G 410243-500MG
77633-068EA 1318 USD
77633-068 77659-186 77642-624 77646-654
Methyl α,3-dihydroxybenzeneacetate ≥95%
Methyl α,3-dihydroxybenzeneacetate

Precaución: For Research Use Only

Formula: C₉H₁₀O₄
Storage Temperature: Ambient
CAS Number: 90721-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR