(4-Bromophenoxy)acetonitrile ≥97%

Supplier: AOBChem USA

Warning

410060-5G 410060-25G
77634-998EA 60 USD
77634-998 77659-184
(4-Bromophenoxy)acetonitrile ≥97%
(4-Bromophenoxy)acetonitrile

Precaución: For Research Use Only

Formula: C₈H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD00464958
CAS Number: 39489-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR