5-Chloro-2-fluoro-3-isopropoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

40908-5G 40908-1G 40908-25G 40908-500MG
77634-992EA 342 USD
77634-992 77638-590 77659-178 77646-652
5-Chloro-2-fluoro-3-isopropoxyphenylboronic acid ≥95%
5-Chloro-2-fluoro-3-isopropoxyphenylboronic acid

Precaución: For Research Use Only

Formula: 232.444 g/mol
MW: C₉H₁₁BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD20529454
CAS Number: 2096339-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR