(5,6-Difluoropyridin-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

40844-5G 40844-1G 40844-25G 40844-500MG
77633-066EA 884 USD
77633-066 77638-582 77655-604 77650-590
(5,6-Difluoropyridin-3-yl)boronic acid ≥97%
(5,6-Difluoropyridin-3-yl)boronic acid

Precaución: For Research Use Only

Formula: C₅H₄BF₂NO₂
MW: 158.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21609554
CAS Number: 1366482-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR