2-Bromo-6-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

405996-5G 405996-25G
77634-990EA 160 USD
77634-990 77655-600
2-Bromo-6-(trifluoromethyl)aniline ≥97%
2-Bromo-6-(trifluoromethyl)aniline

Precaución: For Research Use Only

Formula: C₇H₅BrF₃N
MW: 240.02
Storage Temperature: Ambient
MDL Number: MFCD08741383
CAS Number: 58458-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR