5-Fluoro-1-methyl-1H-indazole-6-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Borono-5-fluoro-1-methyl-1H-indazole, (5-Fluoro-1-methyl-1H-indazol-6-yl)boronic acid

40595-5G 40595-25G 40595-1G 40595-500MG
77634-988EA 518 USD
77634-988 77655-598 77642-620 77646-646
5-Fluoro-1-methyl-1H-indazole-6-boronic acid ≥97%
5-Fluoro-1-methyl-1H-indazole-6-boronic acid

Precaución: For Research Use Only

Formula: C₈H₈BFN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18089818
CAS Number: 2096331-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR