3-Fluoro-2-methoxy-4-(trimethylsilyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

40463-5G 40463-25G 40463-1G 40463-500MG
77634-982EA 588 USD
77634-982 77655-592 77642-616 77650-580
3-Fluoro-2-methoxy-4-(trimethylsilyl)pyridine ≥95%
3-Fluoro-2-methoxy-4-(trimethylsilyl)pyridine

Precaución: For Research Use Only

Formula: C₉H₁₄FNOSi
Storage Temperature: Ambient
MDL Number: MFCD30080967
CAS Number: 1865128-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR