2-Ethoxy-4-fluoro-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

40151-5G 40151-25G
77634-980EA 166 USD
77634-980 77659-174
2-Ethoxy-4-fluoro-5-methylphenylboronic acid ≥97%
2-Ethoxy-4-fluoro-5-methylphenylboronic acid

Precaución: For Research Use Only

Formula: C₉H₁₂BFO₃
MW: 197.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19684096
CAS Number: 2096337-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR