(3-(2-Methoxyethoxy)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

39833-5G 39833-25G 39833-1G 39833-500MG
77634-978EA 333 USD
77634-978 77659-172 77642-612 77646-644
(3-(2-Methoxyethoxy)phenyl)boronic acid ≥95%
(3-(2-Methoxyethoxy)phenyl)boronic acid

Precaución: For Research Use Only

Formula: C₉H₁₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD11183195
CAS Number: 227305-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR