2-[(4-Bromophenoxy)methyl]-2-methyloxirane ≥97%

Supplier: AOBChem USA

Warning

398186-5G 398186-25G 398186-1G 398186-500MG
77633-058EA 1034 USD
77633-058 77659-170 77642-610 77650-578
2-[(4-Bromophenoxy)methyl]-2-methyloxirane ≥97%
2-[(4-Bromophenoxy)methyl]-2-methyloxirane

Precaución: For Research Use Only

Formula: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
CAS Number: 1318734-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR