3'-Chloro-α,α-dimethyl[1,1'-biphenyl]-4-methanol ≥95%
Supplier: AOBChem USA
|
Warning
|
396370-5G
396370-1G
396370-25G
396370-500MG
77629-088EA
1484.23
USD
77629-088
77638-578
77655-584
77646-638
3'-Chloro-α,α-dimethyl[1,1'-biphenyl]-4-methanol ≥95%
3'-Chloro-α,α-dimethyl[1,1'-biphenyl]-4-methanol
Precaución: For Research Use Only
Formula:
246.73 g/mol MW: C₁₅H₁₅ClO Storage Temperature: Ambient |
CAS Number:
92553-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...