2-Cyclopropylmethoxy-6-methoxyphenylboronic acid pinacol ester ≥95%
Supplier: AOBChem USA
|
Warning
|
39562-5G
39562-1G
39562-25G
39562-500MG
77634-972EA
560
USD
77634-972
77638-576
77655-582
77650-570
2-Cyclopropylmethoxy-6-methoxyphenylboronic acid pinacol ester ≥95%
2-Cyclopropylmethoxy-6-methoxyphenylboronic acid pinacol ester
Precaución: For Research Use Only
Formula:
C₁₇H₂₅BO₄ MW: 304.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13152001 CAS Number: 1204580-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...