2-(1H-Pyrazol-1-yl)pyridine-5-boronic acid, pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
39552-5G
39552-25G
39552-1G
39552-500MG
77633-052EA
855
USD
77633-052
77659-160
77642-594
77646-636
2-(1H-Pyrazol-1-yl)pyridine-5-boronic acid, pinacol ester ≥97%
2-(1H-Pyrazol-1-yl)pyridine-5-boronic acid, pinacol ester
Precaución: For Research Use Only
Formula:
C₁₄H₁₈BN₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD11878223 CAS Number: 1218791-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...