4-(Cyclopropylmethoxy)-1-iodo-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

395289-5G 395289-1G 395289-25G 395289-500MG
77633-050EA 998 USD
77633-050 77638-574 77659-158 77650-566
4-(Cyclopropylmethoxy)-1-iodo-2-methylbenzene ≥95%
4-(Cyclopropylmethoxy)-1-iodo-2-methylbenzene

Precaución: For Research Use Only

Formula: C₁₁H₁₃IO
Storage Temperature: Ambient
CAS Number: 1369920-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR