1-(4-(Cyclopropylmethoxy)-2-methylphenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
395288-5G
395288-25G
395288-1G
395288-500MG
77629-086EA
1484.23
USD
77629-086
77659-156
77642-592
77646-634
1-(4-(Cyclopropylmethoxy)-2-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(4-(Cyclopropylmethoxy)-2-methylphenyl)-2,2,2-trifluoroethanone
Precaución: For Research Use Only
Formula:
258.236 g/mol MW: C₁₃H₁₃F₃O₂ Storage Temperature: Ambient |
CAS Number:
3031812-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...