Ethyl 2-(2-isopropoxy-5-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

395223-5G 395223-25G 395223-1G 395223-500MG
77629-082EA 1484.23 USD
77629-082 77635-768 77638-570 77646-630
Ethyl 2-(2-isopropoxy-5-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-isopropoxy-5-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 304.262 g/mol
MW: C₁₄H₁₅F₃O₄
Storage Temperature: Ambient
CAS Number: 3031792-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR