4-(4-Methoxyphenyl)-1-butene ≥95%

Supplier: AOBChem USA

Warning

394844-5G 394844-25G 394844-1G 394844-500MG
77633-048EA 920 USD
77633-048 77655-576 77642-588 77650-564
4-(4-Methoxyphenyl)-1-butene ≥95%
4-(4-Methoxyphenyl)-1-butene

Precaución: For Research Use Only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07698650
CAS Number: 20574-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR