(3-Isopropoxy-4-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

394513-5G 394513-25G 394513-1G 394513-500MG
77633-046EA 1441 USD
77633-046 77659-152 77642-584 77646-626
(3-Isopropoxy-4-methylphenyl)(methyl)sulfane ≥95%
(3-Isopropoxy-4-methylphenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 196.309 g/mol
MW: C₁₁H₁₆OS
Storage Temperature: Ambient
CAS Number: 3031784-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR