1-(5-Bromo-3-ethyl-2-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

393289-5G 393289-25G 393289-1G 393289-500MG
77629-076EA 1484.23 USD
77629-076 77655-570 77642-582 77646-622
1-(5-Bromo-3-ethyl-2-fluorophenyl)ethanol ≥95%
1-(5-Bromo-3-ethyl-2-fluorophenyl)ethanol

Precaución: For Research Use Only

Formula: 247.1040 g/mol
MW: C₁₀H₁₂BrFO
Storage Temperature: Ambient
CAS Number: 3037696-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR