2-(5-Bromo-3-ethyl-2-fluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

392658-5G 392658-1G 392658-25G 392658-500MG
77629-074EA 1484.23 USD
77629-074 77638-566 77659-150 77646-620
2-(5-Bromo-3-ethyl-2-fluorophenyl)propan-2-ol ≥95%
2-(5-Bromo-3-ethyl-2-fluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₄BrFO
Storage Temperature: Ambient
CAS Number: 3037703-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR