2-(3-Ethoxy-2,6-difluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

392339-5G 392339-1G 392339-25G 392339-500MG
77629-072EA 1484.23 USD
77629-072 77638-562 77655-566 77650-560
2-(3-Ethoxy-2,6-difluorophenyl)propan-2-ol ≥95%
2-(3-Ethoxy-2,6-difluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 216.224 g/mol
MW: C₁₁H₁₄F₂O₂
Storage Temperature: Ambient
CAS Number: 3031787-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR