1-(3-Ethoxy-2,6-difluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

392337-5G 392337-1G 392337-500MG 392337-25G
77629-070EA 1484.23 USD
77629-070 77638-560 77652-762 77655-564
1-(3-Ethoxy-2,6-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-Ethoxy-2,6-difluorophenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 254.153 g/mol
MW: C₁₀H₇F₅O₂
Storage Temperature: Ambient
CAS Number: 3031808-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR