1-(4-Ethoxy-2,3-difluorophenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

392011-5G 392011-1G 392011-500MG 392011-25G
77629-062EA 1484.23 USD
77629-062 77638-552 77652-758 77655-560
1-(4-Ethoxy-2,3-difluorophenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(4-Ethoxy-2,3-difluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For Research Use Only

Formula: C₁₃H₁₈F₂O₂
MW: 244.278 g/mol
Storage Temperature: Ambient
CAS Number: 3037676-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR