(4-Ethoxy-2,3-difluorophenyl)triethylsilane ≥97%

Supplier: AOBChem USA

Warning

391997-5G 391997-25G 391997-1G 391997-500MG
77633-036EA 1441 USD
77633-036 77659-138 77642-570 77646-604
(4-Ethoxy-2,3-difluorophenyl)triethylsilane ≥97%
(4-Ethoxy-2,3-difluorophenyl)triethylsilane

Precaución: For Research Use Only

Formula: 272.406 g/mol
MW: C₁₄H₂₂F₂OSi
Storage Temperature: Ambient
CAS Number: 3037703-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR