1-(4-Ethoxy-2,3-difluorophenyl)butan-1-one ≥97%

Supplier: AOBChem USA

Warning

391995-5G 391995-25G 391995-1G 391995-500MG
77629-054EA 1484.23 USD
77629-054 77659-136 77642-568 77650-556
1-(4-Ethoxy-2,3-difluorophenyl)butan-1-one ≥97%
1-(4-Ethoxy-2,3-difluorophenyl)butan-1-one

Precaución: For Research Use Only

Formula: 228.235 g/mol
MW: C₁₂H₁₄F₂O₂
Storage Temperature: Ambient
CAS Number: 3037704-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR