1-(4-Ethoxy-2,3-difluorophenyl)prop-2-en-1-ol ≥97%

Supplier: AOBChem USA

Warning

391993-5G 391993-1G 391993-25G 391993-500MG
77629-052EA 1484.23 USD
77629-052 77638-548 77659-134 77646-602
1-(4-Ethoxy-2,3-difluorophenyl)prop-2-en-1-ol ≥97%
1-(4-Ethoxy-2,3-difluorophenyl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 214.209 g/mol
MW: C₁₁H₁₂F₂O₂
Storage Temperature: Ambient
CAS Number: 3037705-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR