2-(4-Ethoxy-2,3-difluorophenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

391991-1G 391991-5G 391991-500MG
77638-546EA 657 USD
77638-546 77655-554 77650-554
2-(4-Ethoxy-2,3-difluorophenyl)propan-2-ol ≥97%
2-(4-Ethoxy-2,3-difluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 216.224 g/mol
MW: C₁₁H₁₄F₂O₂
Storage Temperature: Ambient
CAS Number: 1781521-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR