1-(4-Chloro-2,6-dimethylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

388922-5G 388922-1G 388922-500MG
77629-046EA 1484.23 USD
77629-046 77642-560 77646-594
1-(4-Chloro-2,6-dimethylphenyl)propan-1-ol ≥95%
1-(4-Chloro-2,6-dimethylphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₅ClO
Storage Temperature: Ambient
CAS Number: 1034980-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR