3-Bromo-2,4-dichloro-6-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

388799-5G 388799-25G 388799-1G 388799-500MG
77629-040EA 1573.84 USD
77629-040 77659-126 77642-558 77650-548
3-Bromo-2,4-dichloro-6-methoxyphenol ≥97%
3-Bromo-2,4-dichloro-6-methoxyphenol

Precaución: For Research Use Only

Formula: C₇H₅BrCl₂O₂
Storage Temperature: Ambient
CAS Number: 3031799-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR