1-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

388628-5G 388628-25G 388628-1G 388628-500MG
77629-036EA 1484.23 USD
77629-036 77655-548 77642-556 77646-592
1-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)propan-1-one ≥95%
1-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: 266.255 g/mol
MW: C₁₁H₁₀F₄OS
Storage Temperature: Ambient
CAS Number: 3031808-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR