Cyclopropyl(2,6-dichloro-3-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

388545-5G 388545-25G 388545-1G 388545-500MG
77629-034EA 1484.23 USD
77629-034 77635-760 77642-554 77650-544
Cyclopropyl(2,6-dichloro-3-methoxyphenyl)methanol ≥95%
Cyclopropyl(2,6-dichloro-3-methoxyphenyl)methanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂Cl₂O₂
Storage Temperature: Ambient
CAS Number: 3037695-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR