4-(Propylthio)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
38847-5G
38847-25G
38847-500MG
77634-970EA
166
USD
77634-970
77655-546
77895-806
4-(Propylthio)phenylboronic acid ≥97%
4-(Propylthio)phenylboronic acid
Precaución: For Research Use Only
Formula:
C₉H₁₃BO₂S Storage Temperature: Ambient |
MDL Number:
MFCD06801730 CAS Number: 279261-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...