α-[2-[(1,1-Dimethylethyl)dimethylsilyl]ethynyl]cyclohexanemethanol ≥95%

Supplier: AOBChem USA

Warning

387122-5G 387122-25G 387122-1G 387122-500MG
77629-030EA 1484.23 USD
77629-030 77635-758 77642-550 77646-588
α-[2-[(1,1-Dimethylethyl)dimethylsilyl]ethynyl]cyclohexanemethanol ≥95%
α-[2-[(1,1-Dimethylethyl)dimethylsilyl]ethynyl]cyclohexanemethanol

Precaución: For Research Use Only

Formula: 252.47 g/mol
MW: C₁₅H₂₈OSi
Storage Temperature: Ambient
CAS Number: 887586-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR