5-Bromo-4-chloro-2-methyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

38709-5G 38709-1G 38709-25G 38709-500MG
77634-966EA 268 USD
77634-966 77638-536 77659-124 77650-542
5-Bromo-4-chloro-2-methyl-2H-indazole ≥97%
5-Bromo-4-chloro-2-methyl-2H-indazole

Precaución: For Research Use Only

Formula: 245.504 g/mol
MW: C₈H₆BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD33403792
CAS Number: 2375917-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR