3-[(1,1-Dimethylethyl)dimethylsilyl]-1-phenyl-2-propyn-1-one ≥95%

Supplier: AOBChem USA

Warning

386998-5G 386998-1G 386998-25G 386998-500MG
77629-028EA 1484.23 USD
77629-028 77638-534 77659-122 77646-586
3-[(1,1-Dimethylethyl)dimethylsilyl]-1-phenyl-2-propyn-1-one ≥95%
3-[(1,1-Dimethylethyl)dimethylsilyl]-1-phenyl-2-propyn-1-one

Precaución: For Research Use Only

Formula: 244.4 g/mol
MW: C₁₅H₂₀OSi
Storage Temperature: Ambient
CAS Number: 849941-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR