1-[(1,1-Dimethylethyl)dimethylsilyl]-1-pentyn-3-ol ≥97%

Supplier: AOBChem USA

Warning

386872-5G 386872-25G 386872-1G 386872-500MG
77629-026EA 1484.23 USD
77629-026 77659-120 77642-548 77646-584
1-[(1,1-Dimethylethyl)dimethylsilyl]-1-pentyn-3-ol ≥97%
1-[(1,1-Dimethylethyl)dimethylsilyl]-1-pentyn-3-ol

Precaución: For Research Use Only

Formula: 198.38 g/mol
MW: C₁₁H₂₂OSi
Storage Temperature: Ambient
CAS Number: 1823666-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR